Adapter, Septum Inlet, Ace Glass Incorporated
Supplier: Ace Glass
5113-13 5113-23
89082-704EA 107.84 CAD
89082-704 89082-706
Adapter, Septum Inlet, Ace Glass Incorporated
Adapters Adapters, Joints with Tube Connectors
This is a sampling adapter with a thread at top that accepts a cap with hole and a PTFE faced septum to allow insertion of a syringe needle.
  • Bottom [ST] inner joint
  • Threaded top
  • Accommodates syringe needle
  • Cap and septum included
  • Fabricated from borosilicate glass
Adapter features a standard taper inner joint at the bottom and an 8-425 GPI thread at the top that accepts an 89075-244 cap with hole and an 89064-072 PTFE faced septum to allow insertion of a syringe needle. Supplied complete with cap and septum.