Hose Connection Adapter, [SJ], Ace Glass Incorporated
Supplier: Ace Glass
5216-23 5216-35
89051-424EA 80.83 CAD
89051-424 89051-426
Hose Connection Adapter, [SJ], Ace Glass Incorporated
Adapters Adapters, Joints with Tube Connectors
These adapters feature a straight hose connection at the top.
  • Bottom spherical ball joint
  • Top hose connection
  • Fabricated from borosilicate glass
Adapters have a spherical ball joint at the bottom and a straight hose connection at the top.