Hose Connection Adapter, [ST], Ace Glass Incorporated
Supplier: Ace Glass
5216-10 9069-04 9069-06 5216-15 9069-05 5216-16
89051-404EA 76.64 CAD
89051-404 89051-484 89051-488 89051-492 89064-830 10767-892
Hose Connection Adapter, [ST], Ace Glass Incorporated
Adapters Adapters, Joints with Tube Connectors
These adapters feature a straight hose connection at top.
  • Bottom ground [ST] inner joint
  • Top hose connection
  • Fabricated from borosilicate glass
Adapters have a ground standard taper inner joint at the bottom and a straight hose connection at the top.