Adapter, PTFE, Ace Glass Incorporated
Supplier: Ace Glass
5223-13 5223-20 5223-38 5223-58
89051-436EA 423.07 CAD
89051-436 89051-438 89051-440 89051-442
Adapter, PTFE, Ace Glass Incorporated
Adapters Adapters, Joints with Tube Connectors
This adapter is used to connect a vial or a bottle to the vapor tube of a rotary evaporator.
  • One end is [ST] outer joint with rib
  • Female vial or bottle GPI thread on other end
  • Fabricated from PTFE
One end of this adapter is a standard taper outer joint with rib for using clamp, while the other end is a female vial or bottle GPI thread. Joint connects to Thread. Thread size represents the GPI Thread Size.