Adapter, Distilling Trap, Ace Glass Incorporated
Supplier: Ace Glass
5225-15 9086-02 5225-10
89051-446EA 234.57 CAD
89051-446 89064-862 89051-444
Adapter, Distilling Trap, Ace Glass Incorporated
Distillation Components Distillation Adapters
Distilling trap adapters are placed above flask to prevent carryover of reactions that have a tendency to foam.
  • Top [ST] outer joint
  • Bottom [ST] inner joint
  • Fabricated from borosilicate glass
Adapter features a standard taper outer joint at the top and a standard taper inner joint at the bottom.