O-Ring Joint to Standard Taper Inner Joint with Frit Connecting Adapter, Ace Glass Incorporated
Supplier: Ace Glass
5117-18 5117-21
89082-620EA 191.93 CAD
89082-620 89082-622
O-Ring Joint to Standard Taper Inner Joint with Frit Connecting Adapter, Ace Glass Incorporated
Adapters Adapters, Joints with Tube Connectors
This adapter is used with vacuum manifolds as port connections.
  • O-ring joint on one end
  • Glass frit below o-ring joint
  • Inner [ST] joint at opposite end
  • One o-ring included
  • Fabricated from borosilicate glass
Adapter features a #15 o-ring joint on one end with a Porosity B (70-100 micron) glass frit below the o-ring joint. An inner standard taper joint is at the opposite end. Supplied with one o-ring.