Adapter, Moisture Trap, Ace Glass Incorporated
Supplier: Ace Glass
5179-07 9101-20
89051-360EA 324.92 CAD
89051-360 89064-988
Adapter, Moisture Trap, Ace Glass Incorporated
Adapters Adapters, Joints with Tube Connectors
This is a unique adapter used in place of a Dean-Stark moisture test receiver.
  • Top [ST] outer joint
  • Bottom [ST] inner joint
  • Side arm [ST] inner joint
  • Fabricated from borosilicate glass
Simply add a condenser (not included) to the top standard taper outer joint, any graduated funnel from 125mL to 2000mL (not included) to bottom standard taper inner joint, attach a sample flask (not included) to the standard taper inner side arm joint, and you have a moisture test receiver.