Adatper, Vacuum Take-Off, Long Stem, Ace Glass Incorporated
Supplier: Ace Glass
5195-10 5195-15 9124-06
89051-378EA 159.29 CAD
89051-378 89051-380 89074-286
Adatper, Vacuum Take-Off, Long Stem, Ace Glass Incorporated
Adapters Adapters, Joints with Tube Connectors
This adapter is designed for use with distillation set-ups.
  • 105° angle outer joint
  • Long stem
  • With hose connection
  • Fabricated from borosilicate glass
Featuring an outer joint that is at an angle of 105°. The hose connection is on the side opposite of the upper joint. Length values represent the stem length below bottom joint (mm).
Order Now
SPECIFICATIONS
Environmentally Preferable