Thermal Transfer Printable Labels, Brady
Supplier: Brady
62371 62366 62363 62359 62379 62354 62365 62355
CA80090-268EA 569.42 CAD
CA80090-268 CA80090-258 CA80090-270 CA80090-262 CA80090-260 CA80090-266 CA80090-256 CA80090-264
Thermal Transfer Printable Labels, Brady
Labels
Printable, self-laminating polyester labels for thermal transfer printers.
  • Good print smudge resistance
  • Solvent resistance
  • Good high and low temperature performance
B-461 performs well in common laboratory environments such as liquid nitrogen and autoclave applications. Used for laboratory identification such as vials, centrifuge tubes, test tubes, straws, and slides. It can be printed with Series R6200 ribbon.
The B-461 self laminating label is for 1 to 2 ml cryo vials-short tail, slit in liner for easy removal. It can be offered in a self-laminating format which has a white thermal printable zone and a clear overlaminating area.
Note: Testing described in this TDS was performed on materials printed with the R4300 Series ribbon.
These labels can be used with, Brady 2461, Brady 3481, Brady 6441, Brady 300MVP Plus, Brady 200MVP Plus, Brady 300X-Plus II, Brady 360X-Plus II, Brady 600X-Plus II, Tagus T200, Tagus T300, Thermal transfer printers, Brady IP, BBP81 printers.
Certifications: B-461 is RoHS compliant to 2005/618/EC MCV amendment to RoHS Directive 2002/95/EC.