Thermal Transfer Printer Labels, Brady
Supplier: Brady Worldwide
11263 11264 11265 11266 11267
CA80090-242EA 241.86 CAD
CA80090-242 CA80090-244 CA80090-246 CA80090-248 CA80090-250
Thermal Transfer Printer Labels, Brady
Labels
These Freezerbondz™ printable labels are ideal for thermal transfer printers. Freezerbondz™ labels can be applied to frozen surfaces including glass and polypropylene stored in liquid nitrogen.
  • Good print smudge resistance, solvent resistance
  • Good high and low temperature performance
Freezerbondz™ labels perform well in common laboratory environments such as liquid nitrogen and autoclave applications. For tube/vial applications, these labels must be wrapped upon itself with at least ¹/₈" overlap.
For use wuth Brady 2461, Brady 3481, Brady 6441, Brady 300MVP Plus, Brady 200MVP Plus, Brady 300X-Plus II, Brady 360X-Plus II, Brady 600X-Plus II, Tagus T200, Tagus T300, Thermal transfer printers, Brady IP and BBP81 printers.