Bulk TLS 2200®/TLS PC Link Labels, B-461, Brady Worldwide®
Supplier: BRADY CANADA CA
BPTL-11-498 62650
CA80090-220EA 406.99 CAD
80090-220 CA80090-220
Bulk TLS 2200®/TLS PC Link Labels, B-461, Brady Worldwide®
The BMP71 label maker can print on more than 30 different types of label materials - all designed with Brady's industrial-strength durability
Bulk roll of labels for the TLS 2200® and TLS-PC Link™ Thermal Transfer Portable Printers. BMP71 Labels white B-498 repositionable vinyl cloth. UL recognized, RoHS compliant. Repositionable rubber based adhesive. B-498 has a specially formulated top coat for very good thermal transfer print quality. The adhesive and cloth backing give excellent holding power, yet allow for clean removal and repositioning. B-498 is used for wire, cable, and general identification numbers, letters and conduit and voltage markers. Thermal transfer print technology. Coated vinyl cloth material type. White, yellow and orange. Repositionable rubber based adhesive. B-498 is a UL recognized component to UL969 labeling and marking standard when printed with Brady series R6200 ribbon. Brady B-498 is RoHS compliant to 2005/618/EC MCV amendment to RoHS Directive 2002/95/EC. Maximum characters per line for font no. 2: 11. Maximum lines of text for font no. 2: 8.
Brady's laboratory labels are ideal identification solutions for the biotechnology, agricultural, environmental and forensic research industry. These lab labels are designed to "match" existing vial, slide, and well plate sizes, and they hold up under tissue processing protocols and other extreme conditions. Brady's laboratory labels are clearly readable by anyone in your lab, and allow for easy bar coding and scanning - which drastically reduces errors. Halogen-Free (DIN VDE 0472/parts 815). Permanent acrylic adhesive. Freezer, liquid nitrogen, autoclave, hot water baths (100 degree celsius) environment. Vial/Tube Size is 1.0 - 8.0ml Vial/Tube. B-461 can be offered in a self-laminating format which has a white thermal printable zone and a clear overlaminating area. B-461 has good print smudge resistance, solvent resistance, good high and low temperature performance. B-461 performs well in common laboratory environments such as liquid nitrogen and autoclave applications. Thermal transfer print technology. Clear film with matte white printable zone coated ink finish. Brady B-461 is RoHS compliant to 2005/618/EC MCV amendment to RoHS Directive 2002/95/EC. Maximum characters per line for font no. 0.1. Maximum lines of text for font no. 0.0902777777777778