Polyester Labels for Thermal Printing, Brady
Supplier: Brady Worldwide
30127 30221 30235 62374
CA80090-284EA 99.38 CAD
CA80090-284 CA80090-286 CA80090-288 CA80090-294
Polyester Labels for Thermal Printing, Brady
Labels
BradyBondz™ polyester labels for thermal transfer printers.
  • Glossy white finish with permanent acrylic
  • Temperature range: −70 to 110 °C
Permanent Polyester (B-423) is ideal for electronic PCB and component identification, bar code labels, rating plates and solar panel identification.
B-423 is suitable for outdoor use on glass, thermoset polyester plastic and polyvinyl fluoride plastic surfaces to support solar panel identification applications.
Certifications: RoHS compliant to 2005/618/EC MCV, UL, CSA & AGA approved with its respective ribbon.
Meets requirements of a halogen-free material per DIN VDE 0472 part 815.