PC Link Labels, Brady
Supplier: Avantor
CA80090-154EA 128.96 CAD
CA80090-154 CA80090-156
PC Link Labels, Brady
Labels
B-439 is a colored vinyl film with an acrylic pressure sensitive adhesive and a topcoat specifically formulated for thermal transfer printing.
Vinyl colors include the following: Silver, Gold, Red, Purple, Yellow, Orange, Green, Light Blue, Black, and White. B-439 is designed for use in ambient conditions with limited solvent exposure. Recommended ribbons are Brady Series R4900 and R6000 black ribbons and R4400W white ribbon.