Adapter, Adjustable Electrode, Ace-Thred, Ace Glass Incorporated
Supplier: Ace Glass
5037-03 5037-10 5037-08 5037-12 5037-20 5037-202 5037-22 5037-30
89051-210EA 88.77 CAD
89051-210 89051-212 89051-214 89051-216 89051-218 89051-220 89051-222 89051-224
Adapter, Adjustable Electrode, Ace-Thred, Ace Glass Incorporated
Adapters Adapters, Conical Ground Joints
This Ace-Thred adapter is used with platinum or other wire electrodes that normally would be step sealed to glass.
  • Electrode may be adjusted or removed
  • Threaded bushing
  • Silicone rubber septum
  • Fabricated from borosilicate glass
With threaded bushing and silicone rubber septum for sealing, the electrode may be adjusted for depth or removed if desired. Glass capillary is 1-1/2 to 2mm with 76mm of stem for sealing to apparatus. Size is determined by bore of hole in bushing. Over length of glass is 140mm. Bushing Size represent the value of Bushing HoleMore Product Information