Slips, Trips, and Falls Signs, National Marker
Supplier: ACCUFORM MANUFACTURING, INC
C609AB C609PB C609RB C628AB C628PB C628RB C643AB C643PB C643RB C658AB C658PB C658RB CBA6P CBA6R CBA7P CBA7R CGA12AB CGA12AP CGA12P CGA12PB CGA12R CGA12RB CGA14P CGA14PB CGA14R CGA14RB CGA31AB CGA31PB CGA31RB CGA34AB CGA34PB CGA34RB D623AB D623PB D623RB ESC203PB ESC203PC ESC203RB ESC203RC ESC366AB ESC366PB ESC366RB ESC37PB ESC37PC ESC37RB ESC37RC ESC57PB ESC57PC ESC57RB ESC57RC N363AB N363PB N363RB
94007-096EA 94.42 CAD
94007-096 94007-098 94007-100 94007-214 94007-216 94007-218 94007-298 94007-300 94007-302 94007-304 94007-306 94007-308 94007-394 94007-396 94007-398 94007-654 94007-656 94007-658 94007-660 94007-722 94007-724 94007-726 94007-728 94007-730 94007-732 94007-734 94007-736 94007-738 94007-740 94007-826 94007-828 94007-830 94007-844 94007-846 94007-848 94009-708 94009-710 94009-712 94009-714 94011-284 94011-286 94011-288 94013-124 94013-126 94013-128 94013-130 94013-172 94013-174 94013-176 94013-178 94013-180 94013-182 94013-184 94013-238 94013-240 94013-242 94013-244 94018-846 94018-848 94018-850 94023-404 94023-406 94023-408
Slips, Trips, and Falls Signs, National Marker
Signs
Prevent slips, trips and falls in your facility by alerting employees and visitors to potential hazards.
  • Ideal for indoor and outdoor labeling
  • Available in Alum, PS vinyl and Rigid plastic (thick polystyrene) materials
  • .040 alum is weatherproof, non-corrosive and has baked-on enamel on both sides
  • .004 PS vinyl material has pressure sensitive adhesive backing
  • .050 rigid plastic material has radius corners and pre-drilled corner mounting holes