Adapter, Straight Connecting, Borosilicate Glass, Ace Glass Incorporated
Supplier: Ace Glass
5035-05 5035-10 5035-15 5035-25 5035-30 5035-35
89051-196EA 111.03 CAD
89051-196 89051-198 89051-200 89051-202 89051-204 89051-206
Adapter, Straight Connecting, Borosilicate Glass, Ace Glass Incorporated
Adapters Adapters, Joints with Tube Connectors
This straight connecting adapter comes with either standard taper or spherical joints at both the top and the bottom.
  • Inner joint on one end and outer joint on other
  • Standard taper or spherical joints
  • Fabricated from borosilicate glass
The length stated is the approximate overall length. Full-sized outer standard taper joints are reinforced.