Adapter, Straight Connecting, Ace Glass Incorporated
Supplier: Ace Glass
5039-07 5039-13 5039-03 5039-09 5039-11
89082-498EA 138.2 CAD
89082-498 89082-502 89082-564 89082-566 89082-596 89082-600
Adapter, Straight Connecting, Ace Glass Incorporated
Adapters Adapters, Joints with Tube Connectors
This straight connecting adapter has ground standard taper inner joints at both ends.
  • Top ground [ST] inner joint
  • Bottom ground [ST] inner joint
  • Fabricated from borosilicate glass
Adapter is available in various lengths.