Adapter, 75 Degree Angle, Ace Glass Incorporated
Supplier: Ace Glass
5070-15 9052-08 9052-12 5070-10
89051-274EA 140.14 CAD
89051-274 89064-796 89064-798 89051-318
Adapter, 75 Degree Angle, Ace Glass Incorporated
Adapters Adapters, Joints with Tube Connectors
This adapter is bent at a 75° angle.
  • [ST] inner joints at both ends
  • Fabricated from borosilicate glass
Adapter has standard taper inner joints at both ends.