Adapter, 105 Degree Angle, Ace Glass Incorporated
Supplier: Ace Glass
5075-15 5075-35 5075-45 5075-10 9055-04
89051-298EA 149.66 CAD
89051-298 89051-300 89051-302 89051-348 89064-800
Adapter, 105 Degree Angle, Ace Glass Incorporated
Adapters Adapters, Joints with Tube Connectors
This 105° angle adapter has standard taper joints or spherical joints.
  • 105° angle
  • Top [ST] reinforced outer or socket joint
  • Bottom [ST] inner or ball joint
  • Fabricated from borosilicate glass
Adapter has standard taper joints (outer reinforced at top, inner at bottom) or spherical joints (socket at top, ball at bottom).