Brushes, Cleanroom Safe, Excelta Corp®
Supplier: Excelta
213A-N 213B-N 213C-N 213D-N
94024-188EA 43.78 CAD
94024-188 94024-190 94024-192 94024-194
Brushes, Cleanroom Safe, Excelta Corp®
Brushes
The 'Series 213' cleanroom safe brush has stainless steel handles and new Statigo9 fiber bristles.
  • Designed for cleaning in tight areas
  • Stainless steel handle, corrosion resistant
The bristles are mechanically clamped into the handle for maximum fiber retention. The fibers are similar to nylon; they have a surface resistivity of 10⁸ ohms/sq.