Brushes, Heat Resistant, Excelta Corp®
Supplier: Excelta
212A-H 212B-H 212C-H 212D-H
94024-180EA 42.36 CAD
94024-180 94024-182 94024-184 94024-186
Brushes, Heat Resistant, Excelta Corp®
Brushes
These cleaning brushes are static safe (10⁸-10¹¹ ohms/sq) and are heat-resistant to 370 °F.
  • Stainless steel handles, corrosion resistant
  • Static-safe
They feature stainless steel handles and horse hair bristles which are mechanically clamped into the handle for maximum fiber retention.