Four Star™ Instrument Cleaning Brushes, EXCELTA®
Supplier: Excelta
210B-H 210B-N 210B-P 210B-S 210S-H 210S-N 210S-P 210S-S
94024-156EA 67.83 CAD
94024-156 94024-158 94024-160 94024-162 94024-164 94024-166 94024-168 94024-170
Four Star™ Instrument Cleaning Brushes, EXCELTA®
Brushes
These instrument cleaning brushes are designed for general cleaning in tight areas.
  • Handle is .25" in diameter
  • Tip size of .13" wide and .19" long
The fibers are mechanically clamped into the stainless steel tube and inserted in the acetal handle. These brushes are used for scratch-free dry brushing.