Adapter, Offset, #7 Ace-Thred, Ace Glass Incorporated
Supplier: Ace Glass
5032-18 5032-22 5032-25
89051-188EA 137.52 CAD
89051-188 89051-190 89051-192
Adapter, Offset, #7 Ace-Thred, Ace Glass Incorporated
Adapters Adapters, Conical Ground Joints
This thermometer adapter comes with an inner standard taper joint at bottom and top threaded piece offset and angled approximately 10° for use in multiple neck flasks.
  • Offset and angled 10°
  • Threaded top
  • Bottom inner [ST] joint
  • Bushing and o-ring included
  • Fabricated from borosilicate glass
Threaded nylon bushing tightens into the glass piece to form an o-ring compression seal with thermometers, bleed tubes, etc. up to 7mm diameter. The thread at top allows for variable vertical positioning of thermometers. And because of the 10° angle, by rotating the joint, you can position the thermometer in the bottom of the flask. Supplied complete with nylon bushing and FETFE o-ring.
Ordering information: For extra o-rings, order 89060-958. For extra bushings, order 89051-122.