Adapter, Ace-Thred, Ace Glass Incorporated
Supplier: Ace Glass
5030-04 5030-06 5030-08 5030-16 5030-19 5030-20 5030-22 5030-24 5030-28 5030-40 5030-42 5030-44 5030-45 5030-60
89051-144EA 95.62 CAD
89051-144 89051-146 89051-148 89051-150 89051-152 89051-154 89051-156 89051-158 89051-160 89051-162 89051-164 89051-166 89051-168 89051-174
Adapter, Ace-Thred, Ace Glass Incorporated
Adapters Adapters, Conical Ground Joints
This adapter is available in both "Midi" and "Maxi" formats
  • "Midi" format is #11 Ace-Thred
  • "Maxi" format is #15 Ace-Thred
  • Bottom ground [ST] inner or spherical ball joint
  • Top Ace-Thred joint
  • Suitable for most vacuum work
  • Fabricated from borosilicate glass
The #11 Ace-Thred sizes will accept inner tubes with diameters of 9mm to 10.5mm, such as thermowells, gas dispersion tubes, vacuum takeoffs; the #15 Ace-Thred sizes will accept inner tubes with diameters of 12.5mm to 14mm such as electrodes, inlet and outlet tubes, etc. Suitable for most vacuum work. Available as complete item, with nylon bushing and FETFE o-ring, or as glassware only.
It comes with a ground standard taper inner or spherical ball joint at the bottom and an Ace-Thred joint at the top.
Ordering information: For extra FETFE o-rings, order 89060-966 (for #11 Ace-Thred) or 89060-984 (for #15 Ace-Thred). For extra bushings, order 89059-214 (for #11 Ace-Thred) or 89059-220 (for #15 Ace-Thred).