Adapter, "Giant" #25 or #36 Ace-Thred, Ace Glass Incorporated
Supplier: Ace Glass
5030-52 5030-55 5030-70 5030-76 5030-78 8067-18 8067-20 8067-22 8067-24 5030-84 5030-80 5030-86 5030-90 5030-88
89051-170EA 122.28 CAD
89051-170 89051-172 89051-176 89051-178 89051-180 89061-902 89061-904 89061-906 89082-540 89082-582 89082-584 89082-586 89082-588 89082-602
Adapter, "Giant" #25 or #36 Ace-Thred, Ace Glass Incorporated
Adapters Adapters, Conical Ground Joints
This "Giant" adapter comes with a ground inner standard taper joint at the bottom and an Ace-Thred at top.
  • Bottom ground inner standard taper joint
  • Either #25 or #36 Ace-Thred at top
  • Can be used with ultrasonic equipment
  • Fabricated from borosilicate glass
Adapter features either a #25 or #36 Ace-Thred at the top that accepts outside diameters of 24-25mm and 34-35mm
89051-176 will accept a 89075-570 slide adapter; 89051-17 will accept a 89075-572 slide adapter. Available as complete item, with nylon bushing and FETFE O-ring, or as glassware only.
(Note: Joint size limits size O.D. of inserted tube.)
Ordering information: For extra FETFE O-rings, order 89061-016 (size -212 for #25 Ace-Thred) or 89061-026 (size -217 for #36 Ace-Thred). For extra bushings, order 89059-226 (for #25 Ace-Thred) or 89059-228 (for #36 Ace-Thred).