Adapter, Straight Connecting with Easy Action Stopcock, Ace Glass Incorporated
Supplier: Ace Glass
3845-05 3845-10
89082-730EA 380.81 CAD
89082-730 89082-732
Adapter, Straight Connecting with Easy Action Stopcock, Ace Glass Incorporated
Adapters Adapters, Conical Ground Joints
This adapter has an Easy Action Stopcock and o-ring joints at top and bottom.
  • Easy Action Stopcock
  • O-ring joints at top and bottom
  • Two o-rings included
  • Fabricated from borosilicate glass
Adapter features an Easy Action Stopcock and #15 o-ring joints at top and bottom.
O-rings are made of FETFE. FETFE is an exclusive compound which is a fluoroelastomer with TFE additives. It has good compression set along with excellent temperature and chemical compatibility. USP Class VI material.
Ordering information: Comes complete with two 89061-002 FETFE o-rings.