Adapter, Tube, "Mini" #7 Ace-Thred, Ace Glass Incorporated
Supplier: Ace Glass
5027-05 5027-20
89051-092EA 43.92 CAD
89051-092 89051-094
Adapter, Tube, "Mini" #7 Ace-Thred, Ace Glass Incorporated
Adapters Adapters, Conical Ground Joints
This adapter is a straight tube with Ace-Thred at one end for use with a nylon bushing.
  • Straight tube
  • Medium wall
  • Fabricated from borosilicate glass
This adapter is a straight tube with a #7 Ace-Thred at one end for use with a nylon bushing. The outside diameter of the tube is 12.7mm, medium wall. The length is approximately 114mm.