Glow Visi Signs™, National Marker
Supplier: ACCUFORM MANUFACTURING, INC
GLV1 GLV28 GLV3 GLV30 GLV31 GLV33 GLV40 GLV41 GLV42 GLV44 GLV46 GLV50 GLV52 GLV53
94015-008EA 156.29 CAD
94015-008 94015-010 94015-014 94015-016 94015-018 94015-022 94015-024 94015-026 94015-028 94015-030 94015-032 94015-034 94015-036 94015-038 94015-040 94021-696
Glow Visi Signs™, National Marker
Signs
An alternative to flat signs, Visi-Signs™ stand out from the wall, allowing for viewing from multiple directions
Made from durable 3mm thick Ultra White PVC. Projects from wall at 45⁰ angle. Features pre-drilled mounting holes for fast and easy installation. Excellent fire properties with a UL 94 V-0 flammability rating.