Tweezers, Broad Flat Wafer Handling, Excelta Corp®
Supplier: Excelta
25-SA 35A-SA 35A-SA-SE 36A-SA
63041-684EA 222.18 CAD
63041-684 63042-024 63042-026 63042-040
Tweezers, Broad Flat Wafer Handling, Excelta Corp®
Forceps
These are ideal for picking flat objects off a flat surface.
  • Large flat tip offers better control of item
  • Anti-magnetic stainless steel, cleanroom safe
  • Two different quality grades
These tweezers feature straight large flat round pointed tips and serrated handles for better control. They are made of anti-magnetic Neverust® stainless steel and is 4.75" long. We offer 2 different qualities. The higher the quality, the more precise the tips.
Order Now
SPECIFICATIONS
Environmentally Preferable