Adapter, Straight Connecting with PTFE Stopcock, Borosilicate Glass, Ace Glass Incorporated
Supplier: Ace Glass
3843-09 3843-33 3843-11 3843-37 3843-55 3843-15 3843-40 3843-58
89082-718EA 532.36 CAD
89082-718 89082-720 89082-750 89082-752 89082-762 89082-780 89082-782 89082-800
Adapter, Straight Connecting with PTFE Stopcock, Borosilicate Glass, Ace Glass Incorporated
Adapters Adapters, Joints with Tube Connectors
This straight connecting adapter has a PTFE stopcock plug.
  • PTFE stopcock plug
  • Available with [ST] inner and reinforced outer joints or spherical ball and socket joints
  • Fabricated from borosilicate glass
Adapter features a 1:5 PTFE stopcock plug and standard taper inner and reinforced outer joints, or spherical ball and sockets joint, at top and bottom.