Tweezer, Ceramic Straight Fine Tip, Excelta Corp®
Supplier: Excelta
277-CE-ET 277-CE 149B-CE 149B-CE-ET 749A-CE 749A-CE-ET
63041-770EA 527.63 CAD
63041-770 75880-570 76471-530 76471-532 76471-580 76471-582
Tweezer, Ceramic Straight Fine Tip, Excelta Corp®
Forceps
Ceramic tweezers offer both heat resistant (2000 °F) for high temp applications and rigidity.
  • Available with ergonomic grips
  • Extremely hard tips
The 'Style 277-CE' white Zirconia ceramic tipped tweezer is 5.5" long. The tip size is 0.04" wide and the shank is made of nickel silver, which is non-magnetic. Zirconia ceramic is six times harder than stainless steel, non-magnetic and chemically inert. Solder will not adhere to the surface. They have a working temperature of over 2000 °F. The 'ET' suffix additionally features Ergo-Tweeze® soft cushioned ergonomic grips.
Order Now
SPECIFICATIONS
Environmentally Preferable