Tweezers, Acetal Soft Replaceable Tip, Excelta Corp®
Supplier: Excelta
159C-RTW 159D-RTW
94024-110EA 93.03 CAD
94024-110 94024-114 76471-378 76471-584
Tweezers, Acetal Soft Replaceable Tip, Excelta Corp®
Forceps
The acetal tips are specifically used for handling items that cannot be scratched, like contact lenses.
  • Will not scratch or mar parts
  • Available in several tip widths
  • Cleanroom safe, will not particulate
  • Resistant to most chemicals and solvents
These straight replaceable tip tweezers are 5" long and are used primarily to prevent marring or scratching of delicate parts. Acetal has outstanding resistance to chemicals and solvents. They are electrically insulative and resistant to temperatures up to 185 °F. The bodies are made of anti-magnetic Neverust®More Product Information