PHCbi MDF Series –30 °C Biomedical Upright Freezers, PHC Corporation
Supplier: PHC Corporation
PHCbi
MDFU731MPA MDFU731PA MDFU731PA MDFU731MPA
89429-298EA 17175.17 CAD
89429-298 CA89429-298 CA10067-690 10067-690
PHCbi MDF Series –30 °C Biomedical Upright Freezers, PHC Corporation
Freezers Laboratory / General Purpose Freezers
PHCbi MDF Series biomedical freezers are designed for short- or intermediate-term storage at temperatures as low as –30 °C.
  • For intermediate storage
  • Uniform temperature maintenance
  • Built-in microprocessor
  • Two-year manufacturer's parts and labor warranty
  • Three-year manufacturer's compressor parts warranty
Constructed with high-performance laboratory and clinical-grade refrigeration systems, these freezers are used in medical, biotechnology, and industrial laboratories for storage of blood components, enzymes, culture media, reagents and specimens.
Uniform temperatures are maintained throughout the chamber using shelf freezing construction. The PHCbi-built microprocessor controller, with digital display of all input/output functions, provides an intuitive method to maintain set point, alarm monitoring, and diagnostic functions. PHCbi’s freezing coils within the shelf offer exceptional temperature recovery due to the expanded surface area offered by the shelf freezing design. Rigid polyurethane foam insulation.
Ordering information: All units are supplied with a two-year manufacturer's parts and labor warranty and a three-year manufacturer's compressor parts warranty. Bins and shelves are sold separately.