VWR® Amber HDPE Carboys with Spigots
Supplier: VWR International
89217-230 89217-232 89217-234 89217-236
89217-230EA 433.15 CAD
89217-230 89217-232 89217-234 89217-236
VWR® Amber HDPE Carboys with Spigots
Bottles Carboys
VWR® amber carboys meet USP standards for light-transmission, making them ideal for storing photo-sensitive materials, while providing users with a highly versatile container that maximizes storage efficiency and ease of use.
  • Conserve benchtop space with rectangular shape
The distinctive rectangular shape saves valuable bench space.
VWR® amber carboys are available in HDPE, offering excellent chemical resistance. VWR® amber carboys are available with or without a spigot. Models with a spigot are compatible with versatile Barbs, which create bottom tubulation options in a variety of hose barb sizes. Versatile caps, adapters, versatile Barbs, and spigots are constructed of polypropylene and are autoclavable at 15 psi. and 121 °C (250 °F) for 20 minutes.
Large handles on top of the container and molded handles in the base provide a full, secure grip to make lifting, handling, and pouring easy. The optional 120 mm (4³/₄") wide neck allows users to reach inside the carboy for easier cleaning. Leakproof polypropylene versatile caps offer the ultimate in connection flexibility. Open caps (sold separately) are compatible with interchangeable adapters, which feature a wide variety of top-connections, including molded-in hose barbs and quick-connect hose barbs. Carboy material and volume are clearly imprinted on the outside of the bottle as well as large, easy to read, metric graduation marks which are certified to ±5% accuracy.
Certifications: Compliance with FDA requirements for food use.
Ordering information: Carboys are supplied with a closed cap; open caps are sold separately.