Safety Lockout Steel Interlocking Hasps, NMC
Supplier: ACCUFORM MANUFACTURING, INC
SL1 SL15
94021-300EA 43.88 CAD
94021-300 94021-304
Safety Lockout Steel Interlocking Hasps, NMC
Security Locks
Vinyl coated high-tensile steel features bent tabs on jaw closures to prevent tampering
Accommodates a total of 6 padlocks up to ⅜" shackle diameter. Available in 1" and 1½" diameter jaws.