EcoPolyBlend™ Drum Management Systems, Justrite®
Supplier: Justrite
28666 28667 28668 28669 28671
89134-670EA 2465.96 CAD
89134-670 89134-672 89134-674 89134-676 89134-678
EcoPolyBlend™ Drum Management Systems, Justrite®
Drum Storage
These modules store drums efficiently, contain spills, and allow safe dispensing of liquids.
  • Store drums efficiently, contain spills, and allow safe dispensing of liquids
  • Base modules with oversized sump hold 110L (30gal.) or 200L (55gal.) drums
  • Stack modules double capacity and include nylon strapping for safety
  • Environmentally friendly black modules are made from 100% recycled polyethylene
  • Molded-in lock channels hold drums in place and dispensing shelf reduces the risk of spills
The base modules with oversized sump hold 110L (30gal.) or 200L (55More Product Information