Citrajet® Low-Foaming Liquid Acid Cleaners
Supplier: Alconox
2001 2055 2015
CA47732-996CS 1151.3 CAD
CA47732-996 CA47732-992 CA47732-994
Citrajet® Low-Foaming Liquid Acid Cleaners
Cleaning Agents Detergents and Surface Cleansers
Citrajet is a low foaming, phosphate-free, concentrated, liquid cleaner and metal brightener for high performance removal of metal oxides, scales, salts and inorganic residues in labware washers, parts washers, power-spray systems and manual and ultrasonic cleaning. Corrosion inhibited and free rinsing leaving no interfering residues. Ideal for acid washing and rinsing.
  • Dilute 2:100
  • Ideal for use as acid wash or rinse
  • Concentrated, phosphate free, biodegradable and readily disposable
  • Combines acid and detergent ingredients to remove metal oxides, salts and
More Product Information