VISI SIGNS™, National Marker
Supplier: ACCUFORM MANUFACTURING, INC
VS10W VS10R VS11R VS11W VS13R VS13W VS15R VS15W VS16R VS16W VS1W VS20R VS20W VS27W VS28W VS31W VS33W VS38W VS3R VS3W VS40W VS41W VS42W VS44W VS46W VS4R VS4W VS50W VS51W VS52W VS5R VS5W
89185-978EA 84.72 CAD
89185-978 89185-998 89186-000 89186-002 94022-922 94022-926 94022-928 94022-934 94022-936 94022-938 94022-940 94022-942 94022-944 94022-948 94022-952 94022-954 94022-956 94022-964 94022-966 94022-970 94022-972 94022-974 94022-976 94022-978 94022-980 94022-982 94022-984 94022-986 94022-988 94022-990 94022-992 94022-994 94022-996 94022-998 94023-000 94023-002 94023-004 94023-006 94023-008 94023-010
VISI SIGNS™, National Marker
Signs
An alternative to flat signs, Visi-Signs™ stand out from the wall, allowing for viewing from multiple directions
Made from durable 3mm thick Ultra White PVC . Projects from wall at 45⁰ angle. Features pre-drilled mounting holes for fast and easy installation. Excellent fire properties with a UL 94 V-0 flammability rating.