Technik™ Tapered Head Stainless Steel Cutters, Aven
Supplier: Aven Tools
10326 10325
47743-096EA 112.45 CAD
47743-096 47743-098
Technik™ Tapered Head Stainless Steel Cutters, Aven
Cutters feature box joint construction and dual leaf springs for comfortable action.
  • Made of mirror polished stainless steel
  • ESD-safe grips
Cutting edges are induction hardened.