Nalgene® High Profile Closures with Color Coders for Micro Packaging Vials, Thermo Scientific
Supplier: Thermo Fisher Scientific
362830-0112 362830-0113 362830-0114 362830-0115 362830-0116 362830-0117 362830-0118 362830-0119 362830-0110 362830-0111
73520-806CS 464.52 CAD
73520-806 73520-808 73520-810 73520-812 73520-814 73520-816 73520-818 73520-820 73521-328 73521-330
Nalgene® High Profile Closures with Color Coders for Micro Packaging Vials, Thermo Scientific
Closures Test/Sample Tube Closures
Nalgene Closures with Color Coders are made of high density polyethylene, and have no O-ring to fall out or contaminate contents
The color coders are permanently attached, are free of toxic heavy metals, and don not come in contact with vial contents. These high profile closures are non-cytotoxic and non-pyrogenic.