Nalgene® Boston Bottles, Round, Opaque White, HDPE, without Closures, Bulk Pack, Thermo Scientific
Supplier: Thermo Fisher Scientific
362008-0032 362008-0016 362008-0008 362008-0004 362008-0002 362008-0001
13500-128CS 476.53 CAD
13500-128 13500-130 13500-132 13500-134 13500-136 16210-880
Nalgene® Boston Bottles, Round, Opaque White, HDPE, without Closures, Bulk Pack, Thermo Scientific
Bottles Plastic Bottles
Nalgene boston round opaque white HDPE bottles are specifically designed for demanding lab use, and have durable uniform walls that are resistant to splitting or puncturing.
They are reliable, durable, chemical resistant, and are opaque white for light sensitive products. They also meet the requirements of light-resistant containers per USP latest edition.