81-Well Cryogenic Storage Racks with Lid, Thermo Scientific
Supplier: Thermo Scientific
8810 8800
53225-618CS 252 CAD
53225-618 53225-620
81-Well Cryogenic Storage Racks with Lid, Thermo Scientific
Racks Cryogenic Vial Racks
Thermo Scientific Molecular BioProducts 81-Well Cryogenic Storage Rack holds eighty-one 1.5 or 2.0mL cryogenic storage vials
They are molded with an alphanumeric grid for easy tube identification. The lid on the tube protects samples and allows racks to be stacked.