Flipper Racks, 4-Way, Thermo Scientific
Supplier: Thermo Scientific
8850 8860 8870
53225-626CS 1414.78 CAD
53225-626 53225-628 53225-630
Flipper Racks, 4-Way, Thermo Scientific
Racks Tube Racks
Thermo Scientific Molecular BioProducts 4-Way Flipper Rack holds thirty-two 0.5mL, thirty-two 1.5mL, twelve 15mL or four 50mL tubes, each on a different side.