Nalgene® Rectangular Clearboys, Thermo Scientific
Supplier: Thermo Fisher Scientific
DS2213-0020 DS2213-0050
16115-307EA 446.74 CAD
16115-307 16115-329
Nalgene® Rectangular Clearboys, Thermo Scientific
Bottles Carboys
Nalgene® rectangular clearboys have a space-saving design for storing media, solutions and sterile water.
These clearboys feature a wide mouth opening for filling and cleaning, as well as a stainless-steel handle which makes it easy to transport. Molded in graduations in 1 gallon and 5 L increments make it easy to dispense liquids. Additionally, the clearboys come with a polypropylene screw closure and TPE gasket.