Personal Protection (PPE), OSHA Danger Signs, National Marker
Supplier: National Marker
D468P D201R D424AB D616PB D474AB D624RB D604PB D201RB D630RB D574RB D429AB D627RB D630PB D166RB D313R D627AB D616RB D238R D468PB D525PB D523RB D464RB D414RB D201A D628PB D469PB D414PB D424PB D464P D526PB D424RB D502PB D238RB D201PB D201P D526AB D525AB D629AB D424R D201EB D574PB D494RB D469AB D414R D502RB D627PB D626PB D604RB D429R D502AB D629PB D468AB D624PB D526RB D604AB D628AB D474RB D626RB D630AB D526EB D524AB D429P D464R D469R D626AB D523PB D414P D494PB D523AB D624AB D238P D469RB D429PB D469P D464AB D424P D384AB D494AB D166PB D524RB D574AB D313P D616AB D201AB D424EB D313RB D464PB D631PB D524PB D525RB D631AB D468R D628RB D631RB D429RB D468RB D238PB D474PB D629RB D384P
94008-562EA 43.2 CAD
94008-562 94009-786 94009-784 94009-782 94008-560 94009-788 94011-244 94011-242 94009-790 94011-240 94010-406 94009-114 94010-402 94010-404 94009-110 94009-112 94010-632 94010-630 94010-470 94010-472 94010-198 94011-304 94011-302 94010-626 94010-628 89185-848 94010-622 94009-732 94010-468 94009-730 94010-624 89185-844 89185-846 94010-620 89185-840 94011-308 89185-842 94011-306 94009-734 94011-310 94008-766 94008-764 94011-314 94011-312 94010-618 94010-614 94008-768 94010-616 94010-214 94010-610 94010-612 94010-216 94010-210 94010-212 94011-318 94011-316 94011-322 94011-168 94011-320 94011-326 94011-324 94010-608 94010-208 94010-204 94010-206 94009-276 94010-200 94009-274 94010-202 94009-792 94009-838 94010-162 94010-164 94011-328 94010-160 94011-332 94011-330 94011-336 94011-334 94011-170 94011-292 94011-290 94010-956 94011-172 94011-294 94009-842 94010-952 94010-954 94009-840 94010-276 94010-156 94010-158 94010-272 94009-728 89185-852 94010-274 94009-846 89185-850 94009-844 94008-770
Personal Protection (PPE), OSHA Danger Signs, National Marker
Signs
OSHA signs are the perfect way to alert employees and guests when a hazard is present in the facility.
  • Ideal for indoor and outdoor labeling
  • Available in aluminum, PS vinyl, fiberglass and rigid plastic (thick polystyrene) materials
  • 0.040 aluminum is weatherproof, non-corrosive and has baked-on enamel on both sides
  • 0.004 PS vinyl material has pressure sensitive adhesive backing
  • 0.095 fiberglass material is rigid, shatter-proof and dent resistant
  • 0.050 rigid plastic material has radius corners and pre-drilled corner mounting holes
Keep facilities safe by displaying an OSHA regulated sign today, and prevent an accident tomorrow. Each sign header reads "Danger."
Certifications: Comply with OSHA 29 CFR 1910.