Open Case Carts, Lakeside®
Supplier: Lakeside
7910 7910W 7910SS 7910SD 7920 7920W 7920SS 7920SD 7930 7930W 7930SS 7930SD 7940 7940W 7940SS 7940SD
89210-152EA 8215.51 CAD
89210-152 89210-154 89210-156 89210-158 89210-160 89210-162 89210-164 89210-166 89210-168 89210-170 89210-172 89210-174 89210-176 89210-178 89210-180 89210-182
Open Case Carts, Lakeside®
Trolleys
Open case carts can be configured to meet your every need
Maximum design flexibility for your best fit. Multiple uses as case carts, supply carts, distribution and linen carts. Stainless steel frame is easy to clean and sanitize. Solid stainless steel or wire shelves allow options for different size supplies. All-welded construction eliminates nuts and bolts for a stronger structure and easier to clean surface. Fits elevators and adaptable for cart pick-up systems. 6" casters for easy maneuverability. Easy to configure with 51/2" space between shelf rungs. No tools required for assembly.