Alpha-Bungarotoxin, CF® Dye and Other Conjugates, Biotium
Supplier: Biotium
00005 00006 00007 00009 00010-1 00011 00012 00013 00014 00015 00016 00017 00002 00003 00004 00018 00026
89138-070EA 890.12 CAD
89138-070 89138-072 89138-074 89138-078 89138-082 89138-084 89138-086 89138-088 89138-090 89138-092 89138-094 89138-096 71003-982 71004-006 71004-058 71004-296 10018-138
Alpha-Bungarotoxin, CF® Dye and Other Conjugates, Biotium
Protein Linking and Labeling Systems
Conjugates of alpha-bungarotoxin labeled with a selection of our CF® dyes and other labels. Labeled alpha-bungarotoxin conjugates can be used for staining nicotinic acetylcholine receptors at neuromuscular junctions in tissue sections.
  • Choose from nine bright and stable CF® Dye colors or other labels
  • Dye options for super-resolution and 2-photon imaging
A variety of alpha-bungarotoxin CF® Dye conjugates offers advantages in brightness, photostability, and/or signal-to-noise ratio compared to other fluorescent dyes. Also available alpha-bungarotoxin conjugates with biotin, fluorescein (FITC), tetramethylrhodamine (TRITC), or sulforhodamine-101 (Texas Red®) and unconjugated alpha-bungarotoxin.
CF® dyes are covered by pending U.S. and international patents. CF® is a trademark of Biotium, Inc. Texas Red® is a registered trademark of Molecular Probes, Inc.