Finnpipette® F2 GLP Kits, Thermo Scientific
Supplier: Thermo Fisher Scientific
4700870 4700885 4700880
89096-132KT 2802.9 CAD
89096-132 89096-136 89096-134
Finnpipette® F2 GLP Kits, Thermo Scientific
Pipettes Micropipettes
Thermo Scientific Finnpipette® F2 GLP Kits provide durable, high-performance Finnpipette® F2 pipetters in an all-inclusive kit containing everything needed for pipetting.
The Thermo Scientific Finnpipette® F2 Good Laboratory Pipetting kits offer the maximum reliability and comfort of the new Finnpipette® F2. The F2 pipetters have unique features such as AVG (Advanced Volume Gearing mechanism) and a very large display for improved usability and comfort.
Ordering information: In addition to the range of Finnpipette® F2 pipettors listed below, each kit includes F-Stand, F2 Pen, F2More Product Information