Molecular BioProducts Low Retention Pipette Tips, Thermo Scientific
Supplier: Thermo Scientific
3501-05 3500-05-11 3511-05 3512-05 3510-05 3721-05 3751-05 3752-05 3551-05 3771-05
72830-422CS 1826.44 CAD
72830-422 72830-426 72830-428 72830-430 72830-432 72830-446 72830-456 72830-459 72830-460 72830-466
Molecular BioProducts Low Retention Pipette Tips, Thermo Scientific
Pipette Tips
Molecular BioProducts low retention pipette tips are specifically designed for eliminating sample retention within the pipette tip.
All Pure tips are racked, pre-sterilized, and certified free of RNase, DNase, DNA, and pyrogen.
The inner surface of the tip is extremely hydrophobic, reducing loss of sample from adhesion to the tip interior. Tips are crystal clear to allow easy sample viewing.