Precoiled Pipe Markers- Blank Yellow, National Marker
Supplier: ACCUFORM MANUFACTURING, INC
A6002 B6002
94003-524EA 28.11 CAD
94003-524 94004-298
Precoiled Pipe Markers- Blank Yellow, National Marker
Signs
Use on wet, oily and dusty pipes where pressure sensitive vinyl markers won't hold
Can be removed and reapplied in other areas.
Certifications: Meets ANSI A13.1-2007